Please login to your online account to display your discounted pricing

5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD10088DA

 View more versions of this product

Catalog No. CD10088DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 229.09
Quantity 1g
Assay 97%
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 88398-93-0
CAS Min % 96.5
Packaging Amber Glass Bottle
Molecular Formula C5H6Cl2N2O2S
MDL Number MFCD00052536
Assay Percent Range 97%
Chemical Name or Material 5-chloro-1, 3-dimethyl-1H-pyrazole-4-sulfonyl chloride
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-chloro-1, 3-dimethyl-1H-pyrazole-4-sulfonyl chloride
CAS 88398-93-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6Cl2N2O2S
Formula Weight 229.09
MDL Number MFCD00052536
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents