missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

$129.45 - $129.45

Chemical Identifiers

CAS 88398-46-3
Molecular Formula C5H8ClN3O2S
Formula Weight 209.66
MDL Number MFCD00105874
Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50546MD SDS Alfa Aesar™
H50546MD
250mg Each for $129.45
AAH5054603 SDS Alfa Aesar™
H5054603
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

88398-46-3
209.66
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide
C5H8ClN3O2S
MFCD00105874

Specifications

250mg
C5H8ClN3O2S
MFCD00105874
88398-46-3
209.66
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning