Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde, 95%

$146.54 - $637.26

Chemical Identifiers

Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde
CAS 27006-76-4
Molecular Formula C6H7ClN2O
Formula Weight 158.59
MDL Number MFCD00052538
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5047503

 
alfa aesar™
H5047503
1g Each for $146.54

AAH5047506

 
alfa aesar™
H5047506
5g Each for $637.26
Description & Specifications

Specifications

Formula Weight 158.59
Chemical Name or Material 5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxaldehyde
Assay 95%
Molecular Formula C6H7ClN2O
MDL Number MFCD00052538
CAS 27006-76-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.