Please login to your online account to display your discounted pricing

5-chloro-1,3-dimethoxybenzene, 97%, ACROS Organics™

$42.77 - $42.77

Chemical Identifiers

Chemical Name or Material 5-Chloro-1, 3-dimethoxybenzene
CAS 7051-16-3
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H9ClO2
Linear Formula ClC6H3(OCH3)2
Formula Weight 172.61g/mol
MDL Number MFCD00008382
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC197770050

  SDS
thermo fisher scientific
197770050
Glass bottle 5g Each for $42.77
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 33° to 37°C
Color White to yellow or pink
Quantity 5g
Chemical Name or Material 5-Chloro-1, 3-dimethoxybenzene
CAS 7051-16-3
CAS Min % 96.0
CAS Max % 100.0
Physical Form White to yellow or pink crystalline powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H9ClO2
Linear Formula ClC6H3(OCH3)2
Formula Weight 172.61g/mol
MDL Number MFCD00008382
Beilstein 06, III, 4335
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.