Please login to your online account to display your discounted pricing

Alfa Aesar™ (5-Carboxypentyl)triphenylphosphonium bromide, 97%

$44.95 - $44.95

Chemical Identifiers

Chemical Name or Material (5-Carboxypentyl)triphenylphosphonium bromide
CAS 50889-29-7
Molecular Formula HO2C(CH2)5P(C6H5)3Br
Formula Weight 457.36
MDL Number MFCD00055556
Synonym (5-Carboxyamyl)triphenylphosphonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2058803

 
alfa aesar™
B2058803
1g Each for $44.95
Description & Specifications

Specifications

Formula Weight 457.36
Chemical Name or Material (5-Carboxypentyl)triphenylphosphonium bromide
Assay 97%
Molecular Formula HO2C(CH2)5P(C6H5)3Br
MDL Number MFCD00055556
CAS 50889-29-7
Synonym (5-Carboxyamyl)triphenylphosphonium bromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 200° to 202°C
Quantity 1g

Applications
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.

Solubility
Soluble in Chloroform and Methanol.

Notes
Hygroscopic. Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.