Please login to your online account to display your discounted pricing

5-Carboxamidotryptamine maleate, Tocris


5-HT1 agonist; Also has high affinity for 5-ht5a and 5-HT7

Quantity 10mg

Manufacturer: r & d systems  045810

 View more versions of this product

Catalog No. 045810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 319.32
Assay Percent Range >99%
Chemical Name or Material 5-Carboxamidotryptamine maleate
Molecular Formula C11H13N3O·C4H4O4
CAS 74885-72-6
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material 5-Carboxamidotryptamine maleate
Assay Percent Range >99%
Molecular Formula C11H13N3O·C4H4O4
Formula Weight 319.32
Accessories

Desiccate at +4°C