Please login to your online account to display your discounted pricing

5-Carboxamidotryptamine maleate, Tocris

5-HT1 agonist; Also has high affinity for 5-ht5a and 5-HT7

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material 5-Carboxamidotryptamine maleate
Assay Percent Range >99%
Molecular Formula C11H13N3O·C4H4O4
Formula Weight 319.32
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

045810

 
r & d systems
045810
10mg Each for $239.63

045850

 
r & d systems
045850
50mg Each for $976.15
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 319.32
Assay Percent Range >99%
Chemical Name or Material 5-Carboxamidotryptamine maleate
Molecular Formula C11H13N3O·C4H4O4
CAS 74885-72-6
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C