Please login to your online account to display your discounted pricing

5-Carbethoxyuracil, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Carbethoxyuracil
CAS 28485-17-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O4
Formula Weight 184.15
MDL Number MFCD00057337
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373970250

  SDS
thermo fisher scientific
AC373970250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373970050

  SDS
fisher scientific acros
AC373970050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 184.15
Quantity 25g
Color White
CAS Max % 100.0
Chemical Name or Material 5-Carbethoxyuracil
Assay Percent Range 98%
Molecular Formula C7H8N2O4
MDL Number MFCD00057337
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 28485-17-8
Additional Information May contain up to one mole of crystal water