Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromouridine, 98%

$32.09 - $32.09

Chemical Identifiers

Chemical Name or Material 5-Bromouridine
CAS 957-75-5
Molecular Formula C9H11BrN2O6
Formula Weight 323.1
MDL Number MFCD00006528
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1850701

  SDS
alfa aesar™
A1850701
0.25g Each for $32.09
Description & Specifications

Specifications

Formula Weight 323.1
CAS 957-75-5
Quantity 0.25g
Melting Point 191° to 193°C
Molecular Formula C9H11BrN2O6
Assay 98%
MDL Number MFCD00006528
Chemical Name or Material 5-Bromouridine

Applications
5-Bromouridine is often incorporated into RNA in order to detect immunocytochemically and analyzed by cytometry. It possesses anti-viral activity and inhibits the activity of viruses such as human immunodeficiency virus.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.