Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromotryptamine hydrochloride, 98%

$129.92 - $392.51

Chemical Identifiers

Chemical Name or Material 5-Bromotryptamine hydrochloride
CAS 81868-12-4
Molecular Formula C10H11BrN2·HCl
Formula Weight 275.58
MDL Number MFCD00152022
Beilstein 3728224
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1941803

 
alfa aesar™
L1941803
1g Each for $392.51

AAL19418MD

 
alfa aesar™
L19418MD
250mg Each for $129.92
Description & Specifications

Specifications

Formula Weight 275.58
Chemical Name or Material 5-Bromotryptamine hydrochloride
Assay 98%
MDL Number MFCD00152022
CAS 81868-12-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H11BrN2·HCl
Melting Point ≈274°C
Beilstein 3728224
Quantity 1g
Health Hazard 1 Warning

Applications
5-Bromotryptamine hydrochloride is used as pharmaceutical intermediates.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with light and oxidizing agents. Store in the dark.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.