Please login to your online account to display your discounted pricing

5-Bromothiophene-2-sulfonyl chloride, 97%


Quantity 1g

Manufacturer: fisher scientific company  KM03757DA

 View more versions of this product

Catalog No. KM03757DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 261.55
Quantity 1g
Assay 97%
Molecular Formula C4H2BrClO2S2
Assay Percent Range 97%
Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride
CAS 55854-46-1

Specifications

Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H2BrClO2S2
Formula Weight 261.55