Please login to your online account to display your discounted pricing

5-Bromothiophene-2-sulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H2BrClO2S2
Formula Weight 261.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM03757DA

  SDS
fisher scientific company
KM03757DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 261.55
Quantity 1g
Assay 97%
Molecular Formula C4H2BrClO2S2
Assay Percent Range 97%
Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride
CAS 55854-46-1