Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromothiophene-2-carboxaldoxime, 97%

$78.35 - $289.30

Chemical Identifiers

Chemical Name or Material 5-Bromothiophene-2-carboxaldoxime
CAS 2160-63-6
Molecular Formula C5H4BrNOS
Formula Weight 206.06
MDL Number MFCD00126375
Synonym 5-Bromothiophene-2-carboxaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2496803

  SDS
alfa aesar™
B2496803
1g Each for $78.35

AAB2496806

  SDS
alfa aesar™
B2496806
5g Each for $289.30
Description & Specifications

Specifications

Formula Weight 206.06
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 1 Warning
MDL Number MFCD00126375
Melting Point 132° to 134°C
Quantity 1g
Assay 97%
Chemical Name or Material 5-Bromothiophene-2-carboxaldoxime
Molecular Formula C5H4BrNOS
CAS 2160-63-6
Synonym 5-Bromothiophene-2-carboxaldehyde oxime
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.