missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 5-Bromothiazole, 98%
$43.04 - $370.09
Chemical Identifiers
CAS | 3034-55-7 |
---|---|
Molecular Formula | C3H2BrNS |
Molecular Weight (g/mol) | 164.02 |
MDL Number | MFCD07787394 |
InChI Key | DWUPYMSVAPQXMS-UHFFFAOYSA-N |
Synonym | 2-bromo-1,4-thiazole, 5-bromo thiazole, 5-bromo-thiazole, 5-bromothiazol, 5-bromothiazole, 5-bromthiazole, pubchem2077, thiazole, 5-bromo, thiazole,5-bromo, zlchem 497 |
PubChem CID | 546059 |
IUPAC Name | 5-bromo-1,3-thiazole |
SMILES | C1=C(SC=N1)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH55387MD
|
Alfa Aesar™
H55387MD |
250mg |
Each for $43.04
|
|
AAH5538703
|
Alfa Aesar™
H5538703 |
1g |
Each for $115.46
|
|
AAH5538706
|
Alfa Aesar™
H5538706 |
5g |
Each for $370.09
|
|
Description
Applications5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
3034-55-7 | |
164.02 | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
546059 | |
C1=C(SC=N1)Br |
C3H2BrNS | |
MFCD07787394 | |
2-bromo-1,4-thiazole, 5-bromo thiazole, 5-bromo-thiazole, 5-bromothiazol, 5-bromothiazole, 5-bromthiazole, pubchem2077, thiazole, 5-bromo, thiazole,5-bromo, zlchem 497 | |
5-bromo-1,3-thiazole |
Specifications
1.835 | |
1.5955 | |
250mg | |
C3H2BrNS | |
2-bromo-1,4-thiazole, 5-bromo thiazole, 5-bromo-thiazole, 5-bromothiazol, 5-bromothiazole, 5-bromthiazole, pubchem2077, thiazole, 5-bromo, thiazole,5-bromo, zlchem 497 | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
5-bromo-1,3-thiazole | |
546059 | |
98% |
76°C (169°F) | |
Moisture sensitive | |
3034-55-7 | |
MFCD07787394 | |
Soluble in water. | |
C1=C(SC=N1)Br | |
164.02 | |
164.02 | |
5-Bromothiazole |
Safety and Handling
GHS H Statement
H301-H318-H227-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H318-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen