Learn More
5-Bromothiazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5538706
Description
5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromothiazole is used as a pharmaceutical intermediate and performance material.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
5-Bromothiazole | |
1.835 | |
C3H2BrNS | |
MFCD07787394 | |
Moisture sensitive | |
Soluble in water. | |
C1=C(SC=N1)Br | |
164.02 | |
164.02 |
3034-55-7 | |
76°C (169°F) | |
1.5955 | |
5 g | |
5-bromothiazole, thiazole, 5-bromo, 5-bromo-thiazole, 5-bromthiazole, thiazole,5-bromo, 5-bromothiazol, 5-bromo thiazole, zlchem 497, pubchem2077, 2-bromo-1,4-thiazole | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
5-bromo-1,3-thiazole | |
546059 | |
98% |
Chemical Identifiers
3034-55-7 | |
164.02 | |
DWUPYMSVAPQXMS-UHFFFAOYSA-N | |
5-bromo-1,3-thiazole |
C3H2BrNS | |
MFCD07787394 | |
546059 | |
C1=C(SC=N1)Br |
Safety and Handling
GHS H Statement
H301-H318-H227-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H318-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only