Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one, 99%

$112.22 - $390.97

Chemical Identifiers

Chemical Name or Material 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one
CAS 75822-54-7
Molecular Formula C10H8BrNO3
Formula Weight 270.08
MDL Number MFCD00097597
Synonym 5-Bromoisatin 3-(ethylene acetal)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5058203

  SDS
alfa aesar™
H5058203
1g Each for $390.97

AAH50582MD

  SDS
alfa aesar™
H50582MD
250mg Each for $112.22
Description & Specifications

Specifications

Formula Weight 270.08
Chemical Name or Material 5'-Bromospiro[1,3-dioxolane-2,3'-indol]-2'(1'H)-one
Assay 99%
Molecular Formula C10H8BrNO3
MDL Number MFCD00097597
CAS 75822-54-7
Synonym 5-Bromoisatin 3-(ethylene acetal)
Quantity 1g
RTECS Number WH1306502