missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromosalicylic acid, 98%, ACROS Organics™

$111.31 - $111.31

Chemical Identifiers

CAS 89-55-4
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00002455
InChI Key IEJOONSLOGAXNO-UHFFFAOYSA-N
Synonym 2-hydroxy-5-bromobenzoic acid, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromosalicyclic acid, 5-bromosalicylic acid, 5-bromsalicylic acid, acmc-209r1n, benzoic acid, 3-bromo-6-hydroxy, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo
PubChem CID 6972
IUPAC Name 5-bromo-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162140500
SDS View Documents
Fisher Scientific Acros
162140500
50g Glass bottle
Each for $111.31
 
AC162142500
SDS View Documents
Thermo Fisher Scientific
162142500
250g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

89-55-4
217.02
IEJOONSLOGAXNO-UHFFFAOYSA-N
6972
OC(=O)C1=CC(Br)=CC=C1O
C7H5BrO3
MFCD00002455
2-hydroxy-5-bromobenzoic acid, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromosalicyclic acid, 5-bromosalicylic acid, 5-bromsalicylic acid, acmc-209r1n, benzoic acid, 3-bromo-6-hydroxy, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo
5-bromo-2-hydroxybenzoic acid

Specifications

Authentic
1% max. (K.F.)
162.0°C to 166.0°C
89-55-4
100.0
C7H5BrO3
MFCD00002455
2-hydroxy-5-bromobenzoic acid, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromosalicyclic acid, 5-bromosalicylic acid, 5-bromsalicylic acid, acmc-209r1n, benzoic acid, 3-bromo-6-hydroxy, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo
OC(=O)C1=CC(Br)=CC=C1O
217.02
217.02
98%
Glass bottle
White to Beige
50g
97.5
98%
BrC6H3(OH)CO2H
10, 107
IEJOONSLOGAXNO-UHFFFAOYSA-N
5-bromo-2-hydroxybenzoic acid
6972
Powder
5-Bromosalicylic acid, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-917-0

RTECSNumber : VO1850000

TSCA : TSCA

SDS
Product Certifications