missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromosalicylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 162140500

 View more versions of this product

Catalog No. AC162140500


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromosalicylic acid
89-55-4
100.0
White to Beige
98%
C7H5BrO3
MFCD00002455
10, 107
IEJOONSLOGAXNO-UHFFFAOYSA-N
5-bromo-2-hydroxybenzoic acid
6972
98%
98%
97.5
162.0°C to 166.0°C
Authentic
Glass bottle
BrC6H3(OH)CO2H
50 g
5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
OC(=O)C1=CC(Br)=CC=C1O
217.02
217.02
Powder

Chemical Identifiers

89-55-4
217.02
IEJOONSLOGAXNO-UHFFFAOYSA-N
6972
OC(=O)C1=CC(Br)=CC=C1O
C7H5BrO3
MFCD00002455
5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
5-bromo-2-hydroxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-917-0

RTECSNumber : VO1850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only