Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromosalicylic acid, 98%

$61.78 - $233.55

Chemical Identifiers

Chemical Name or Material 5-Bromosalicylic acid
CAS 89-55-4
Molecular Formula C7H5BrO3
Formula Weight 217.02
MDL Number MFCD00002455
Beilstein 2209121
Synonym 5-Bromo-2-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2033818

  SDS
alfa aesar™
B2033818
50g Each for $61.78

AAB2033822

  SDS
alfa aesar™
B2033822
100g Each for $112.86

AAB2033830

  SDS
alfa aesar™
B2033830
250g Each for $233.55
Description & Specifications

Specifications

Formula Weight 217.02
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 166° to 170°C
Chemical Name or Material 5-Bromosalicylic acid
MDL Number MFCD00002455
Synonym 5-Bromo-2-hydroxybenzoic acid
Assay 98%
CAS 89-55-4
TSCA TSCA
Quantity 50g
Health Hazard 1 Warning
RTECS Number VO1850000
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H5BrO3
Beilstein 2209121

Applications
5-Bromosalicylic acid is used as organic synthesis. The cross-coupling reaction of 5-bromosalicylic acid with alkyl and aryl Grignard reagents provides a navel efficient synthesis of substituted salicylic acids.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.