Learn More
5-Bromosalicylaldehyde, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 159910250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Bromosalicylaldehyde | |
1761-61-1 | |
100.0 | |
Authentic | |
Glass bottle | |
BrC6H3(OH)CHO | |
25g | |
5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
5-bromo-2-hydroxybenzaldehyde | |
72863 | |
99% |
99% | |
98.5 | |
110°C | |
99% | |
C7H5BrO2 | |
MFCD00003330 | |
08, 54 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
C1=CC(=C(C=C1Br)C=O)O | |
201.02 | |
201.02 |
Chemical Identifiers
1761-61-1 | |
201.02 | |
MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
72863 | |
C1=CC(=C(C=C1Br)C=O)O |
C7H5BrO2 | |
MFCD00003330 | |
5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
5-bromo-2-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 217-167-2
TSCA : TSCA
RUO â Research Use Only