Please login to your online account to display your discounted pricing

5-Bromopyrimidine, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC204301000

 View more versions of this product

Catalog No. AC204301000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Chunks
Formula Weight 158.99
Quantity 100g
Color Yellow to beige-brown
TSCA TSCA
EINECS Number 224-992-1
Melting Point 69° to 73°C
CAS Max % 100.0
Chemical Name or Material 5-Bromopyrimidine
Name Note 98%
Assay Percent Range 98%
Linear Formula (C5H3N2)Br
CAS 4595-59-9
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Packaging Glass bottle
Molecular Formula C4H3BrN2
MDL Number MFCD00006117
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromopyrimidine
Name Note 98%
CAS 4595-59-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H3BrN2
Linear Formula (C5H3N2)Br
Formula Weight 158.99
MDL Number MFCD00006117
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents