Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromopyridine-2-carbonyl chloride, tech. 90%

Chemical Identifiers

Chemical Name or Material 5-Bromopyridine-2-carbonyl chloride
CAS 137178-88-2
Purity Grade Technical Grade
Formula Weight 220.45
MDL Number MFCD07368290
UN Number UN3261
Synonym 5-Bromopicolinoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6176006

  SDS
alfa aesar™
H6176006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6176014

  SDS
alfa aesar™
H6176014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.45
MDL Number MFCD07368290
Sensitivity Moisture Sensitive
Physical Form Solid
Synonym 5-Bromopicolinoyl chloride
Chemical Name or Material 5-Bromopyridine-2-carbonyl chloride
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
UN Number UN3261
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
CAS 137178-88-2
Quantity 5g
Assay 90%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Storage Note 1 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C