missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromopentyl acetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1554922
Description
5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-âBromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications
5-Bromopentyl acetate | |
1.255 | |
>110°C (230°F) | |
1.462 | |
100 g | |
5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester | |
JIHJLQWUVZUKCH-UHFFFAOYSA-N | |
5-bromopentyl acetate | |
85140 | |
98% |
15848-22-3 | |
109°C to 110°C (15 mmHg) | |
C7H13BrO2 | |
MFCD00039204 | |
1752109 | |
Slightly soluble in water. | |
CC(=O)OCCCCCBr | |
209.08 | |
209.09 |
Chemical Identifiers
15848-22-3 | |
209.08 | |
JIHJLQWUVZUKCH-UHFFFAOYSA-N | |
85140 | |
CC(=O)OCCCCCBr |
C7H13BrO2 | |
MFCD00039204 | |
5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester | |
5-bromopentyl acetate |
Safety and Handling
EINECSNumber : 239-961-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only