missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromopentyl acetate, 98%

Manufacturer:  Alfa Aesar™ A1554906

 View more versions of this product

Catalog No. AAA1554906



Description

Description

Applications
5-​Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications

Specifications

5-Bromopentyl acetate
109°C to 110°C (15mmHg)
15848-22-3
MFCD00039204
5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
CC(=O)OCCCCCBr
209.083
209.09
>110°C (230°F)
5g
C7H13BrO2
1752109
JIHJLQWUVZUKCH-UHFFFAOYSA-N
5-bromopentyl acetate
85140
98%

Chemical Identifiers

15848-22-3
209.083
JIHJLQWUVZUKCH-UHFFFAOYSA-N
85140
CC(=O)OCCCCCBr
C7H13BrO2
MFCD00039204
5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
5-bromopentyl acetate
Documents