Please login to your online account to display your discounted pricing

5-Bromooxindole, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  S05248DA

 View more versions of this product

Catalog No. S05248DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.05
Quantity 1g
Assay 97%
Assay Percent Range 97%
Synonym 5-Bromo-1, 3-dihydro-2H-indol-2-one
Chemical Name or Material 5-Bromo-1, 3-dihydro-2H-indol-2-one
Molecular Formula C8H6BrNO
CAS 20870-78-4
Health Hazard 1 Warning

Specifications

Chemical Name or Material 5-Bromo-1, 3-dihydro-2H-indol-2-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6BrNO
Formula Weight 212.05
Synonym 5-Bromo-1, 3-dihydro-2H-indol-2-one
Accessories

Warning