Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromonicotinoyl chloride, 98%

$105.58 - $105.58

Chemical Identifiers

Chemical Name or Material 5-Bromonicotinoyl chloride
CAS 39620-02-5
Molecular Formula C6H3BrClNO
Formula Weight 220.45
MDL Number MFCD00173853
UN Number UN3261
Beilstein 115810
Synonym 5-Bromopyridine-3-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1551006

 
alfa aesar™
L1551006
5g Each for $105.58

AAL1551014

 
alfa aesar™
L1551014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.45
Chemical Name or Material 5-Bromonicotinoyl chloride
Assay 98%
Molecular Formula C6H3BrClNO
MDL Number MFCD00173853
CAS 39620-02-5
UN Number UN3261
Melting Point 72° to 74°C
Synonym 5-Bromopyridine-3-carbonyl chloride
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Beilstein 115810
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.