Please login to your online account to display your discounted pricing

5-Bromonicotinic acid, 98%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC211392500

 View more versions of this product

Catalog No. AC211392500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point >180°C
Physical Form Powder
Assay ≥97.5%
Formula Weight 202.01
Quantity 250g
Color White to yellow-gray or light brown
Beilstein 22, 44
CAS Max % 100.0
Synonym 5-Bromo-3-pyridinecarboxylic acid
CAS 20826-04-4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Chemical Name or Material 5-Bromonicotinic acid
Assay Percent Range 98%
Molecular Formula C6H4BrNO2
Linear Formula Br(C5H3N)COOH
MDL Number MFCD00009783
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 5-Bromonicotinic acid
CAS 20826-04-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4BrNO2
Linear Formula Br(C5H3N)COOH
Formula Weight 202.01
MDL Number MFCD00009783
Synonym 5-Bromo-3-pyridinecarboxylic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents