Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromonicotinic acid, 98%

$39.32 - $411.35

Chemical Identifiers

Chemical Name or Material 5-Bromonicotinic acid
CAS 20826-04-4
Molecular Formula C6H4BrNO2
Formula Weight 202.01
MDL Number MFCD00009783
Beilstein 115854
Synonym 5-Bromopyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1358906

  SDS
alfa aesar™
A1358906
5g Each for $39.32

AAA1358914

  SDS
alfa aesar™
A1358914
25g Each for $109.54

AAA1358922

  SDS
alfa aesar™
A1358922
100g Each for $411.35
Description & Specifications

Specifications

Formula Weight 202.01
Melting Point 182° to 186°C
Chemical Name or Material 5-Bromonicotinic acid
Quantity 5g
Assay 98%
Molecular Formula C6H4BrNO2
MDL Number MFCD00009783
CAS 20826-04-4
Beilstein 115854
Synonym 5-Bromopyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QT0868000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.