missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromonicotinamide, 99%, ACROS Organics™

Chemical Identifiers

CAS 28733-43-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H5BrN2O
Formula Weight 201.02
MDL Number MFCD00173919
Chemical Name or Material 5-Bromonicotinamide
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC355950050 SDS Thermo Fisher Scientific
AC355950050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28733-43-9
100.0
C6H5BrN2O
MFCD00173919
≥98.5%
98.5
99%
201.02
5-Bromonicotinamide

Specifications

Powder
5g
218° to 222°C
Glass bottle
28733-43-9
C6H5BrN2O
MFCD00173919
≥98.5%
201.02
Off-white to light pink
100.0
Authentic
98.5
99%
5-Bromonicotinamide

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.