Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromonicotinamide, 98%

$39.49 - $139.07

Chemical Identifiers

Chemical Name or Material 5-Bromonicotinamide
CAS 28733-43-9
Molecular Formula C6H5BrN2O
Formula Weight 201.03
MDL Number MFCD00173919
Synonym 5-Bromopyridine-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1784006

  SDS
alfa aesar™
A1784006
5g Each for $39.49

AAA1784014

  SDS
alfa aesar™
A1784014
25g Each for $139.07
Description & Specifications

Specifications

Formula Weight 201.03
CAS 28733-43-9
Molecular Formula C6H5BrN2O
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00173919
Health Hazard 1 Warning
Synonym 5-Bromopyridine-3-carboxamide
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 226° to 227°C
Chemical Name or Material 5-Bromonicotinamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.