Please login to your online account to display your discounted pricing

5-(bromomethyl)-2-methyl-4-(trifluoromethyl)-1,3-thiazole, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76308DA

 View more versions of this product

Catalog No. CC76308DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 260.08
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 5-(Bromomethyl)-2-methyl-4-(trifluoromethyl)-1, 3-thiazole
CAS 1000339-73-0
MDL Number MFCD09264554
Assay Percent Range 97%
Molecular Formula C6H5BrF3NS
CAS Min % 96.5
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 5-(Bromomethyl)-2-methyl-4-(trifluoromethyl)-1, 3-thiazole
CAS 1000339-73-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H5BrF3NS
Formula Weight 260.08
MDL Number MFCD09264554
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents