missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6240003

 View more versions of this product

Catalog No. AAH6240003



Specifications

Specifications

5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
1256360-47-0
MFCD17926519
UEBRMJPFPXLIMK-UHFFFAOYSA-N
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
53217273
Solid
1g
C13H17BBrFO2
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F
314.989
314.99
96%

Chemical Identifiers

1256360-47-0
314.989
UEBRMJPFPXLIMK-UHFFFAOYSA-N
53217273
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F
C13H17BBrFO2
MFCD17926519
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents