Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
CAS 1256360-47-0
Formula Weight 314.99
MDL Number MFCD17926519
Synonym 5-Bromomethyl-2-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6240003

 
alfa aesar™
H6240003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62400MD

 
alfa aesar™
H62400MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.99
Chemical Name or Material 5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Quantity 1g
Assay 96%
MDL Number MFCD17926519
CAS 1256360-47-0
Synonym 5-Bromomethyl-2-fluorophenylboronic acid pinacol ester
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C