missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H62400MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester | |
C13H17BBrFO2 | |
250 mg | |
UEBRMJPFPXLIMK-UHFFFAOYSA-N | |
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
53217273 | |
96% |
1256360-47-0 | |
MFCD17926519 | |
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-bromomethyl-2-fluorophenylboronic acid pinacol ester, 5-bromomethyl-2-fluorophenylboronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F | |
314.989 | |
314.99 | |
Solid |
Chemical Identifiers
1256360-47-0 | |
314.989 | |
UEBRMJPFPXLIMK-UHFFFAOYSA-N | |
53217273 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F |
C13H17BBrFO2 | |
MFCD17926519 | |
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-bromomethyl-2-fluorophenylboronic acid pinacol ester, 5-bromomethyl-2-fluorophenylboronic acid, pinacol ester | |
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold