missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Bromomethyl)-2,4-diphenyl-1,3-thiazole, Tech., Maybridge

Chemical Identifiers

CAS 876316-44-8
CAS Min % 80.0
CAS Max % 100.0
Molecular Formula C16H12BrNS
Formula Weight 330.25
MDL Number MFCD08271912
Chemical Name or Material 5-(Bromomethyl)-2, 4-diphenyl-1, 3-thiazole
Assay Tech.
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC42308DA SDS Fisher Scientific Company
CC42308DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC42308CB SDS Fisher Scientific Company
CC42308CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

876316-44-8
100.0
330.25
5-(Bromomethyl)-2, 4-diphenyl-1, 3-thiazole
Technical Grade
80.0
C16H12BrNS
MFCD08271912
Tech.

Specifications

330.25
80.0
C16H12BrNS
MFCD08271912
Tech.
1g
100.0
876316-44-8
5-(Bromomethyl)-2, 4-diphenyl-1, 3-thiazole
Technical Grade

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.