Please login to your online account to display your discounted pricing

5-(Bromomethyl)-2,1,3-benzothiadiazole, 95+%

$68.50 - $205.50

Chemical Identifiers

Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5BrN2S
Formula Weight 229.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC09208DA

  SDS
fisher scientific company
CC09208DA
Amber glass bottle 1g Each for $205.50

CC09208EA

 
fisher scientific company
CC09208EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC09208CB

  SDS
fisher scientific company
CC09208CB
Each for $68.50
Description & Specifications

Specifications

Formula Weight 229.1
Quantity 1g
Assay 95%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Assay Percent Range 95%
Molecular Formula C7H5BrN2S
CAS 65858-50-6
Packaging Amber glass bottle