Please login to your online account to display your discounted pricing

5-(Bromomethyl)-2,1,3-benzothiadiazole, 95+%


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC09208EA

 View more versions of this product

Catalog No. CC09208EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.1
Quantity 10g
Assay 95%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Assay Percent Range 95%
Molecular Formula C7H5BrN2S
CAS 65858-50-6
Packaging Amber glass bottle

Specifications

Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5BrN2S
Formula Weight 229.1