missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™
$233.04 - $547.54
Chemical Identifiers
CAS | 499770-76-2 |
---|---|
Molecular Formula | C8H8BrN3 |
Molecular Weight (g/mol) | 226.077 |
InChI Key | OSUZHHPMRAIJDY-UHFFFAOYSA-N |
Synonym | 5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole |
PubChem CID | 2795418 |
IUPAC Name | 5-(bromomethyl)-1-methylbenzotriazole |
SMILES | CN1C2=C(C=C(C=C2)CBr)N=N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC23508CB
|
Thermo Scientific Chemicals
CC23508CB |
250 mg | Amber Glass Bottle |
Each for $233.04
|
|
||||
CC23508DA
|
Thermo Scientific Chemicals
CC23508DA |
1 g | Amber Glass Bottle |
Each for $547.54
|
|
||||
Chemical Identifiers
499770-76-2 | |
226.077 | |
5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole | |
5-(bromomethyl)-1-methylbenzotriazole |
C8H8BrN3 | |
OSUZHHPMRAIJDY-UHFFFAOYSA-N | |
2795418 | |
CN1C2=C(C=C(C=C2)CBr)N=N1 |
Specifications
499770-76-2 | |
Amber Glass Bottle | |
250 mg | |
OSUZHHPMRAIJDY-UHFFFAOYSA-N | |
5-(bromomethyl)-1-methylbenzotriazole | |
2795418 | |
95% |
95% | |
C8H8BrN3 | |
5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole | |
CN1C2=C(C=C(C=C2)CBr)N=N1 | |
226.077 | |
226.08 | |
5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole |