Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromoisoquinoline, 98%

$35.09 - $97.89

Chemical Identifiers

Chemical Name or Material 5-Bromoisoquinoline
CAS 34784-04-8
Molecular Formula C9H6BrN
Formula Weight 208.06
MDL Number MFCD01646405
Beilstein 114468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1963203

 
alfa aesar™
L1963203
1g Each for $97.89

AAL19632MD

 
alfa aesar™
L19632MD
250mg Each for $35.09
Description & Specifications

Specifications

Formula Weight 208.06
Boiling Point 95° to 97°C (0.1mmHg)
Quantity 1g
Assay 98%
Molecular Formula C9H6BrN
MDL Number MFCD01646405
CAS 34784-04-8
Beilstein 114468
Health Hazard 1 Warning
Chemical Name or Material 5-Bromoisoquinoline
Melting Point 81° to 83°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.