missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromoisoquinoline, 95%, Maybridge

Chemical Identifiers

CAS 34784-04-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H6BrN
Formula Weight 208.06
MDL Number MFCD01646405
Synonym 5-Bromoisoquinoline (IUPAC)
Chemical Name or Material 5-Bromoisoquinoline
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC05010DA SDS Fisher Scientific Company
CC05010DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

34784-04-8
100.0
C9H6BrN
MFCD01646405
5-Bromoisoquinoline
94.5
95%
208.06
5-Bromoisoquinoline (IUPAC)
95%

Specifications

208.06
100.0
94.5
5-Bromoisoquinoline (IUPAC)
C9H6BrN
5-Bromoisoquinoline
1g
34784-04-8
Amber Glass Bottle
95%
MFCD01646405
95%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning