Please login to your online account to display your discounted pricing

5-Bromoisophthalic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Bromoisophthalic acid
CAS 23351-91-9
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H5BrO4
Linear Formula BrC6H3(CO2H)2
Formula Weight 245.03
MDL Number MFCD00152019
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC293190010

  SDS
fisher scientific acros
293190010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Powder
Formula Weight 245.03
Quantity 1g
Color White to almost white
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 5-Bromoisophthalic acid
Assay Percent Range 96%
MDL Number MFCD00152019
CAS 23351-91-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Additional Information may contain variable amounts of water
Linear Formula BrC6H3(CO2H)2
Infrared Spectrum Authentic
Molecular Formula C8H5BrO4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.