Please login to your online account to display your discounted pricing

5-Bromoisophthalic acid, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  293190050

 View more versions of this product

Catalog No. AC293190050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Additional Information may contain variable amounts of water
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to almost white
Chemical Name or Material 5-Bromoisophthalic acid
CAS 23351-91-9
CAS Min % 95.0
CAS Max % 100.0
Physical Form White to almost white powder
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C8H5BrO4
Linear Formula BrC6H3(CO2H)2
Formula Weight 245.03g/mol
MDL Number MFCD00152019
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-Bromoisophthalic acid
CAS 23351-91-9
CAS Min % 95.0
CAS Max % 100.0
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C8H5BrO4
Linear Formula BrC6H3(CO2H)2
Formula Weight 245.03g/mol
MDL Number MFCD00152019
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents