Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromoisophthalic acid, 95%

$114.07 - $340.53

Chemical Identifiers

Chemical Name or Material 5-Bromoisophthalic acid
CAS 23351-91-9
Molecular Formula C8H5BrO4
Formula Weight 245.03
MDL Number MFCD00152019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2687406

  SDS
alfa aesar™
H2687406
5g Each for $114.07

AAH2687414

  SDS
alfa aesar™
H2687414
25g Each for $340.53
Description & Specifications

Specifications

Formula Weight 245.03
Melting Point 280° to 286°C
Assay 95%
Odor Odorless
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD00152019
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 23351-91-9
Molecular Formula C8H5BrO4
TSCA TSCA
Quantity 5g
Health Hazard 1 Warning
Chemical Name or Material 5-Bromoisophthalic acid

Applications
5-Bromoisophthalic acid, is used as a pharmaceutical intermediate. It is also used in the polymer industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.