Please login to your online account to display your discounted pricing

5-Bromoisatoic anhydride, 97%


Manufacturer: thermo fisher scientific  AC398570050

 View more versions of this product

Catalog No. AC398570050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 242.02
Quantity 5g
Chemical Name or Material 5-Bromoisatoic anhydride
Molecular Formula C8H4BrNO3
CAS 4692-98-2
Assay Percent Range 97%

Specifications

Chemical Name or Material 5-Bromoisatoic anhydride
CAS 4692-98-2
Molecular Formula C8H4BrNO3
Formula Weight 242.02