Please login to your online account to display your discounted pricing

5-Bromoisatoic anhydride, 97%

Chemical Identifiers

Chemical Name or Material 5-Bromoisatoic anhydride
CAS 4692-98-2
Molecular Formula C8H4BrNO3
Formula Weight 242.02
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AC398570010

  SDS
thermo fisher scientific
AC398570010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398570050

  SDS
thermo fisher scientific
AC398570050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Bromoisatoic anhydride
CAS 4692-98-2
Packaging Glass bottle
Quantity 1g
Assay Percent Range 97%
Molecular Formula C8H4BrNO3
Formula Weight 242.02