Learn More
5-Bromoisatin, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 154230250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Bromoisatin | |
97.5 | |
Authentic | |
Plastic Bottle | |
MFCD00005719 | |
21, 453 | |
MBVCESWADCIXJN-UHFFFAOYSA-N | |
5-bromo-1H-indole-2,3-dione | |
6889 | |
98% |
87-48-9 | |
100.0 | |
97.5% min. (HPLC) | |
C8H4BrNO2 | |
25g | |
5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
C1=CC2=C(C=C1Br)C(=O)C(=O)N2 | |
226.03 | |
226.03 |
Chemical Identifiers
87-48-9 | |
226.03 | |
MBVCESWADCIXJN-UHFFFAOYSA-N | |
6889 | |
C1=CC2=C(C=C1Br)C(=O)C(=O)N2 |
C8H4BrNO2 | |
MFCD00005719 | |
5-bromoisatin, 5-bromoindoline-2,3-dione, 5-bromoindole-2,3-dione, isatin, 5-bromo, 5-bromisatin, 1h-indole-2,3-dione, 5-bromo, 5-bromisatin czech, 5-bromo-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-bromo, 5-bromoisatin monohydrate | |
5-bromo-1H-indole-2,3-dione |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 201-747-7
RUO â Research Use Only