Please login to your online account to display your discounted pricing

5-Bromoindoline, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC398630010

 View more versions of this product

Catalog No. AC398630010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material 5-Bromoindoline
Assay Percent Range 97%
Molecular Formula C8H8BrN
MDL Number MFCD00027410
CAS 22190-33-6

Specifications

Chemical Name or Material 5-Bromoindoline
CAS 22190-33-6
Assay Percent Range 97%
Molecular Formula C8H8BrN
MDL Number MFCD00027410