Please login to your online account to display your discounted pricing

5-Bromoindoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Bromoindoline
CAS 22190-33-6
Assay Percent Range 97%
Molecular Formula C8H8BrN
MDL Number MFCD00027410
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC398630050

  SDS
thermo fisher scientific
AC398630050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398630010

  SDS
thermo fisher scientific
AC398630010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
MDL Number MFCD00027410
CAS 22190-33-6
Molecular Formula C8H8BrN
Chemical Name or Material 5-Bromoindoline
Assay Percent Range 97%