missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromoindole-3-carboxylic acid, 97%

$49.50 - $586.00

Chemical Identifiers

CAS 10406-06-1
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD05664007
InChI Key JVZMBSGNSAHFCY-UHFFFAOYSA-N
Synonym 5-bromoindole-3-carboxylic acid, 5-bromo-indole-3-carboxylic acid, 5-bromo-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid,5-bromo, 1h-indole-3-carboxylic acid, 5-bromo, 5-bromo-1h-indole-3-carboxylicacid, pubchem7982, acmc-1btmm, ksc173q4r, 5-bromo-3-indolecarboxylicacid
PubChem CID 7018243
IUPAC Name 5-bromo-1H-indole-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64834MD View Documents Alfa Aesar™
H64834MD
250mg Each for $49.50
 
AAH6483403 View Documents Alfa Aesar™
H6483403
1g Each for $147.27
 
AAH6483406 View Documents Alfa Aesar™
H6483406
5g Each for $586.00
 
Specifications

Chemical Identifiers

10406-06-1
240.056
JVZMBSGNSAHFCY-UHFFFAOYSA-N
7018243
C1=CC2=C(C=C1Br)C(=CN2)C(=O)O
C9H6BrNO2
MFCD05664007
5-bromoindole-3-carboxylic acid, 5-bromo-indole-3-carboxylic acid, 5-bromo-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid,5-bromo, 1h-indole-3-carboxylic acid, 5-bromo, 5-bromo-1h-indole-3-carboxylicacid, pubchem7982, acmc-1btmm, ksc173q4r, 5-bromo-3-indolecarboxylicacid
5-bromo-1H-indole-3-carboxylic acid

Specifications

250mg
C9H6BrNO2
475219
JVZMBSGNSAHFCY-UHFFFAOYSA-N
5-bromo-1H-indole-3-carboxylic acid
7018243
Solid
5-Bromoindole-3-carboxylic acid
10406-06-1
MFCD05664007
5-bromoindole-3-carboxylic acid, 5-bromo-indole-3-carboxylic acid, 5-bromo-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid,5-bromo, 1h-indole-3-carboxylic acid, 5-bromo, 5-bromo-1h-indole-3-carboxylicacid, pubchem7982, acmc-1btmm, ksc173q4r, 5-bromo-3-indolecarboxylicacid
C1=CC2=C(C=C1Br)C(=CN2)C(=O)O
240.056
240.06
97%
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C