Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Bromoindole-3-carboxaldehyde, 98+%

$33.15 - $33.15

Chemical Identifiers

Chemical Name or Material 5-Bromoindole-3-carboxaldehyde
CAS 877-03-2
Molecular Formula C9H6BrNO
Formula Weight 224.06
MDL Number MFCD00152016
Beilstein 124725
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1907503

  SDS
alfa aesar™
L1907503
1g Each for $33.15
Description & Specifications

Specifications

Formula Weight 224.06
Assay ≥98%
Chemical Name or Material 5-Bromoindole-3-carboxaldehyde
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 877-03-2
Beilstein 124725
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 204° to 206°C
Quantity 1g
Molecular Formula C9H6BrNO
MDL Number MFCD00152016
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.